N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%

Supplier: Thermo Scientific Chemicals
Danger

38240-0010 38240-0050
TS38240-0010EA 251 USD
TS38240-0010 TS38240-0050
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate
Formula: C₃₂H₁₂BF₂₀N
Storage Temperature: Ambient
MDL Number: MFCD12913898
CAS Number: 118612-00-3

Order Now

Specification Test Results

Appearance White to off-white Powder (May have a blue hue)
Infrared spectrum Conforms
NMR (19F-NMR) ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR