Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate

38239-0010 38239-0050
TS38239-0010EA 416 USD
TS38239-0010 TS38239-0050
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Formula: C₄₃H₁₅BF₂₀
MW: 922.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426981
CAS Number: 136040-19-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to dark yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR