2-(Boc-amino)ethanethiol 97%

Supplier: Thermo Scientific Chemicals
Warning

38108-0050 38108-0250
TS38108-0050EA 82 USD
TS38108-0050 TS38108-0250
2-(Boc-amino)ethanethiol 97%
2-(Boc-amino)ethanethiol
Formula: C₇H₁₅NO₂S
Boiling Pt: 68 °C (.3 mmHg)
Density: 1.049 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00274335
CAS Number: 67385-09-5
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4770 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR