Tetrakis(dimethylamino)zirconium(IV)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TDMAZ, Tetrakis(dimethylamino)zirconium

38041-0010 38041-0050
TS38041-0010EA 68 USD
TS38041-0010 TS38041-0050
Tetrakis(dimethylamino)zirconium(IV)
Tetrakis(dimethylamino)zirconium(IV)
Formula: C₈H₂₄N₄Zr
MW: 267.527 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00239502
CAS Number: 19756-04-8
UN: 3396
ADR: 4.3 II

Order Now

Specification Test Results

Appearance Yellow-green Crystalline solid
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR