Tris(2-methoxyphenyl)phosphine 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(o-methoxyphenyl)phosphine

37885-0010 37885-0050
TS37885-0010EA 57 USD
TS37885-0010 TS37885-0050
Tris(2-methoxyphenyl)phosphine 97+%
Tris(2-methoxyphenyl)phosphine
Formula: C₂₁H₂₁O₃P
Melting Pt: 204…208 °C
MDL Number: MFCD00014892
CAS Number: 4731-65-1
EINECS: 225-235-8

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 203 to 208 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR