Triethyl orthobenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha,alpha,alpha-Triethoxytoluene, (Triethoxymethyl)benzene, Orthobenzoic acid triethyl ester

37876-0050 37876-0250
TS37876-0050EA 67 USD
TS37876-0050 TS37876-0250
Triethyl orthobenzoate 97%
Triethyl orthobenzoate
Formula: C₁₃H₂₀O₃
MW: 224.3 g/mol
Flash Pt: 96 °C
MDL Number: MFCD00009222
CAS Number: 1663-61-2
EINECS: 216-771-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4740 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR