1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: SIPr.HCl, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium chloride, 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride

37834-0010 37834-0050
TS37834-0010EA 73 USD
TS37834-0010 TS37834-0050
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 90%
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride
Formula: C₂₇H₃₉ClN₂
MW: 427.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369796
CAS Number: 258278-25-0

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR