1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: SIMes.HCl

37833-0010 37833-0050
TS37833-0010EA 87 USD
TS37833-0010 TS37833-0050
1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride 97%
1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride
Formula: C₂₁H₂₇ClN₂
MW: 342.91 g/mol
Melting Pt: 280…286 °C
Storage Temperature: Ambient
MDL Number: MFCD09039279
CAS Number: 173035-10-4

Order Now

Specification Test Results

Appearance Beige to pink-brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR