Quinaldine red, pure
Supplier: Thermo Scientific Chemicals
37751-0050
37751-0250
37751-1000
TS37751-0050EA
53
USD
TS37751-0050
TS37751-0250
TS37751-1000
Quinaldine red, pure
Quinaldine red
Formula:
C₂₁H₂₃IN₂ Melting Pt: 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011968 CAS Number: 117-92-0 EINECS: 204-221-5 Merck Index: 12,08230 |
Specification Test Results
Appearance | Grey to green-black to dark blue Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...