Methyl-β-cyclodextrin (mixture of several Methylated), M.W. ∼ 1310
Supplier: Thermo Scientific Chemicals
Synonyms:
Methylated-β-Cyclodextrin, Methyl-β-cyclodextrin
37711-0050
37711-0250
37711-1000
TS37711-0050EA
52
USD
TS37711-0050
TS37711-0250
TS37711-1000
Methyl-β-cyclodextrin (mixture of several Methylated), M.W. ∼ 1310
Methyl-beta-cyclodextrin (mixture of several Methylated)
MW:
1303.3000 g/mol Melting Pt: 165…172 °C Flash Pt: 187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074980 CAS Number: 128446-36-6 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5.0 % |
Degree of substitution | 1.7 to 1.9 (NMR) |
Specific optical rotation | +154.0° to +165.0° (20°C, 589 nm) (c=5, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...