4-Nitrobenzyl chloroformate 95%

Supplier: Thermo Scientific Chemicals
Danger

37604-0050 37604-0250
TS37604-0050EA 43 USD
TS37604-0050 TS37604-0250
4-Nitrobenzyl chloroformate 95%
4-Nitrobenzyl chloroformate
Formula: C₈H₆ClNO₄
Melting Pt: 32 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007375
CAS Number: 4457-32-3
EINECS: 224-708-6
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance Off-white to yellow to beige Solid and/or chunks or liquid after melting
Infrared spectrum Conforms
Melting point 30 to 36 °C
Titration Argentometric ≥94 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR