2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 97%

Supplier: Thermo Scientific Chemicals
Danger

37483-0010 37483-0050
TS37483-0010EA 89 USD
TS37483-0010 TS37483-0050
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 97%
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite
Formula: C₉H₁₈ClN₂OP
MW: 236.68 g/mol
Density: 1.061 g/cm³
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00011544
CAS Number: 89992-70-1
UN: 2845
ADR: 4.2,I

Order Now

Specification Test Results

Appearance Clear to hazy colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4800 (20°C, 589 nm)
Chloride (Cl) 14.5 to 15.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR