(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl 2-(triphenylphosphoranylidene)acetate
37062-0010
37062-0050
37062-0250
TS37062-0010EA
32
USD
TS37062-0010
TS37062-0050
TS37062-0250
(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%
(tert-Butoxycarbonylmethylene)triphenylphosphorane
Formula:
C₂₄H₂₅O₂P MW: 376.436 g/mol Melting Pt: 146…151 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075545 CAS Number: 35000-38-5 UN: 3464 ADR: 6.1,II |
Specification Test Results
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 146 to 155 °C |
Carbon (C) | 73.5 to 79.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...