Bis(pyridine)iodonium tetrafluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36968-0010
36968-0050
TS36968-0010EA
61
USD
TS36968-0010
TS36968-0050
Bis(pyridine)iodonium tetrafluoroborate 97%
Bis(pyridine)iodonium tetrafluoroborate
Formula:
C₁₀H₁₀BF₄IN₂ Melting Pt: 137…141 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03703393 CAS Number: 15656-28-7 |
Specification Test Results
Appearance | Light yellow to orange to tan Powder or crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...