tert-Butyl trichloroacetimidate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 2,2,2-trichloroacetimidate, t-Butyl trichioroacetimidate, t-butyl-2,2,2-trichloroacetimidate
36933-0050
36933-0250
36933-1000
TS36933-0050EA
56
USD
TS36933-0050
TS36933-0250
TS36933-1000
tert-Butyl trichloroacetimidate 95%
tert-Butyl trichloroacetimidate
Formula:
C₆H₁₀Cl₃NO MW: 218.502 g/mol Boiling Pt: 64…66 °C (15 mmHg) Melting Pt: 20 °C Density: 1.222 g/cm³ Flash Pt: 55 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077410 CAS Number: 98946-18-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid or low melting crystalline mass |
Infrared spectrum | Conforms |
Refractive index | 1.4550 to 1.4570 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...