N-Methoxy-N-methylacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

36931-0050 36931-0250
TS36931-0050EA 105 USD
TS36931-0050 TS36931-0250
N-Methoxy-N-methylacetamide 98%
N-Methoxy-N-methylacetamide
Formula: C₄H₉NO₂
Boiling Pt: 152 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00060098
CAS Number: 78191-00-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR