4,5-Dicyanoimidazole 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

36853-0050 36853-0250
TS36853-0050EA 79 USD
TS36853-0050 TS36853-0250
4,5-Dicyanoimidazole 99%
4,5-Dicyanoimidazole
Formula: C₅H₂N₄
MW: 118.1 g/mol
Melting Pt: 174…177 °C
MDL Number: MFCD00005194
CAS Number: 1122-28-7
EINECS: 214-344-6

Order Now

Specification Test Results

Appearance White to off-white to light beige Powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR