Tri-tert-butylphosphonium tetrafluoroborate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tri-t-butylphosphonium tetrafluoroborate

36694-0010 36694-0050
TS36694-0010EA 95 USD
TS36694-0010 TS36694-0050
Tri-tert-butylphosphonium tetrafluoroborate 97+%
Tri-tert-butylphosphonium tetrafluoroborate
Formula: C₁₂H₂₈BF₄P
MW: 290.13 g/mol
Melting Pt: 261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04039975
CAS Number: 131274-22-1

Order Now

Specification Test Results

Appearance White Crystals and/or chunks
Infrared spectrum Conforms
NMR (31P-NMR) ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR