Di-tert-butyl azodicarboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Di-tert-butyl diazene-1,2-dicarboxylate
36692-0050
36692-0250
36692-1000
TS36692-0050EA
56
USD
TS36692-0050
TS36692-0250
TS36692-1000
Di-tert-butyl azodicarboxylate 97%
Di-tert-butyl azodicarboxylate
Formula:
C₁₀H₁₈N₂O₄ MW: 230.26 g/mol Melting Pt: 90…92 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00015001 CAS Number: 870-50-8 EINECS: 212-796-9 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 89 to 93 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...