Triphenylphosphine 1,0 - 1,5 mmol/g resin, cross-linked with 1% DVB
Supplier: Thermo Scientific Chemicals
|
Warning
|
36687-0050
36687-0250
TS36687-0050EA
99
USD
TS36687-0050
TS36687-0250
Triphenylphosphine 1,0 - 1,5 mmol/g resin, cross-linked with 1% DVB
Triphenylphosphine
Formula:
C₁₈H₁₅P MW: 262.29 g/mol Boiling Pt: 377 Melting Pt: 79 Flash Pt: 180 Storage Temperature: Refrigerator |
MDL Number:
MFCD00003043 CAS Number: 603-35-0 Merck Index: 12,09873 |
Specification Test Results
Appearance | White to yellow Beads |
Particle size | 100 to 200 mesh |
Crosslinking | 1 % DVB |
Loading | 1 to 1.5 mmol/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...