1,2-Bis(dicyclohexylphosphino)ethane 98%

Supplier: Thermo Scientific Chemicals
Warning

36385-0010 36385-0050
TS36385-0010EA 141 USD
TS36385-0010 TS36385-0050
1,2-Bis(dicyclohexylphosphino)ethane 98%
1,2-Bis(dicyclohexylphosphino)ethane
Formula: C₂₆H₄₈P₂
MW: 422.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015521
CAS Number: 23743-26-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR