Tris(hydroxymethyl)phosphine 95%

Supplier: Thermo Scientific Chemicals
Danger

36384-0010 36384-0250
TS36384-0010EA 79 USD
TS36384-0010 TS36384-0250
Tris(hydroxymethyl)phosphine 95%
Tris(hydroxymethyl)phosphine
Formula: C₃H₉O₃P
MW: 124.076 g/mol
Boiling Pt: 111…113 °C (2.5 mmHg)
Melting Pt: 48…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00055382
CAS Number: 2767-80-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
NMR ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR