Tris[4-(trifluoromethyl)phenyl]phosphine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine

35534-0010 35534-0050
TS35534-0010EA 145 USD
TS35534-0010 TS35534-0050
Tris[4-(trifluoromethyl)phenyl]phosphine 99%
Tris[4-(trifluoromethyl)phenyl]phosphine
Formula: C₂₁H₁₂F₉P
MW: 466.28 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00058883
CAS Number: 13406-29-6

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 75 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR