2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane 92%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-tert-butyl 1,1,4,4-tetramethyltetramethylene diperoxide, Triqanox XQ 8, 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane

34990-0050 34990-1000
TS34990-0050EA 29 USD
TS34990-0050 TS34990-1000
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane 92%
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane
Formula: C₁₆H₃₄O₄
MW: 290.44 g/mol
Boiling Pt: 55…57 °C (7 mmHg)
Melting Pt: 6 °C
Density: 0.87 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00053715
CAS Number: 78-63-7
UN: 3103
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20°C, 589 nm)
GC ≥90 %
Color scale ≤50 APHA
Impurity ≤0.3 % (Hydroperoxides) as 2,5-dihydroperoxy-2,5-dimethyl-hexane

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR