Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(1-methyl-1-phenylethyl)peroxide, Bis(alpha,alpha-dimethylbenzyl)peroxide, Dicumyl Peroxide

34988-1000 34988-5000
TS34988-1000EA 56 USD
TS34988-1000 TS34988-5000
Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) 99%
Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide)
Formula: C₁₈H₂₂O₂
MW: 270.37 g/mol
Melting Pt: 38…41 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036227
CAS Number: 80-43-3
EINECS: 201-279-3
UN: 3110
ADR: 5.2,

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 38 to 41 °C
Titration Iodometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR