Lauroyl peroxide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dilauroyl peroxide

34974-1000 34974-5000
TS34974-1000EA 63 USD
TS34974-1000 TS34974-5000
Lauroyl peroxide 99%
Lauroyl peroxide
Formula: (C₁₁H₂₃CO)₂O₂
MW: 398.628 g/mol
Melting Pt: 53…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008964
CAS Number: 105-74-8
UN: 3106
ADR: 5.2,

Order Now

Specification Test Results

Appearance White Powder or granules or flakes or pellets
Infrared spectrum Conforms
Melting point 53 to 57 °C
Titration Iodometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR