Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Di-μ-hydroxo-bis[(N,N,N'.N'-tetramethylethylenediamine)copper(II)] chloride

34898-0050 34898-0250
TS34898-0050EA 59 USD
TS34898-0050 TS34898-0250
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride 98%
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride
Formula: C₁₂H₃₄Cl₂Cu₂N₄O₂
MW: 464.42 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD01321163
CAS Number: 30698-64-7

Order Now

Specification Test Results

Appearance Dark purple Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR