Toluidine blue O, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride
34860-0050
34860-0250
34860-1000
TS34860-0050EA
50
USD
TS34860-0050
TS34860-0250
TS34860-1000
Toluidine blue O, pure
Toluidine blue O
Formula:
C₁₅H₁₆ClN₃S Storage Temperature: Ambient |
MDL Number:
MFCD00011934 CAS Number: 92-31-9 EINECS: 202-146-2 Merck Index: 12,09658 |
Specification Test Results
Appearance | Dark green Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...