Diisopropyl azodicarboxylate 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

32756-0050 32756-0250 32756-1000 32756-5000
TS32756-0050EA 42 USD
TS32756-0050 TS32756-0250 TS32756-1000 TS32756-5000
Diisopropyl azodicarboxylate 94%
Diisopropyl azodicarboxylate
Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
Boiling Pt: 75 °C (.2 mmHg)
Melting Pt: 3…5 °C
Density: 1.02 g/cm³
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008875
CAS Number: 2446-83-5
EINECS: 219-502-8
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear to hazy yellow to orange to red Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4220 (20°C, 589 nm)
GC ≥92 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR