Bis(diisopropylamino)chlorophosphine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(di-i-propylamino)chlorophosphine

32156-0010 32156-0050 32156-0250
TS32156-0010EA 28 USD
TS32156-0010 TS32156-0050 TS32156-0250
Bis(diisopropylamino)chlorophosphine
Bis(diisopropylamino)chlorophosphine
Formula: C₁₂H₂₈ClN₂P
MW: 266.79 g/mol
Melting Pt: 100…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061482
CAS Number: 56183-63-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 100 to 110 °C
Additional info May contain varying amount of diisopropylamine.HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR