Chloro(di-tert-butyl)phosphine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-tert-butylchlorophosphine, Di-t-butylchlorophosphine
32090-0050
32090-0250
32090-1000
TS32090-0050EA
118
USD
TS32090-0050
TS32090-0250
TS32090-1000
Chloro(di-tert-butyl)phosphine 96%
Chloro(di-tert-butyl)phosphine
Formula:
C₈H₁₈ClP MW: 180.66 g/mol Boiling Pt: 69…70 °C (10 mmHg) Melting Pt: 2…3 °C Density: 0.95 g/cm³ Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008815 CAS Number: 13716-10-4 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...