1,3-Bis(diphenylphosphino)propane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

31005-0050 31005-0250 31005-1000
TS31005-0050EA 75 USD
TS31005-0050 TS31005-0250 TS31005-1000
1,3-Bis(diphenylphosphino)propane 97%
1,3-Bis(diphenylphosphino)propane
Formula: C₂₇H₂₆P₂
Melting Pt: 57…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003050
CAS Number: 6737-42-4
EINECS: 229-791-2

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or granules
Infrared spectrum Conforms
Melting point 57 to 63 °C
Elemental Analysis ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR