(Triphenylphosphoranylidene)acetaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane

30749-0050 30749-0250 30749-1000
TS30749-0050EA 55 USD
TS30749-0050 TS30749-0250 TS30749-1000
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde
Formula: C₂₀H₁₇OP
MW: 304.33 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006994
CAS Number: 2136-75-6

Order Now

Specification Test Results

Appearance Cream to pink to orange to brown Crystalline powder
Infrared spectrum Conforms
Water ≤4.0 % (K.F.)
Titration with HClO4 96.0 to 104.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR