Rhodamine B 98+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

29657-0010 29657-0100 29657-0250 29657-1000
TS29657-0010EA 341 USD
TS29657-0010 TS29657-0100 TS29657-0250 TS29657-1000
Rhodamine B 98+%, pure
Rhodamine B
Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Melting Pt: 210…211 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011931
CAS Number: 81-88-9
EINECS: 201-383-9
Merck Index: 13,08266

Order Now

Specification Test Results

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
HPLC ≥98.0 %
Moisture ≤2.0 % (105°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR