Tri(2-furyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tri(furan-2-yl)phosphine, Tri-2-furylphosphine

29480-0010 29480-0050
TS29480-0010EA 92 USD
TS29480-0010 TS29480-0050 TS29480-2500
Tri(2-furyl)phosphine 98%
Tri(2-furyl)phosphine
Formula: C₁₂H₉O₃P
MW: 232.17 g/mol
Boiling Pt: 136 °C (4 mmHg)
Melting Pt: 61…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151857
CAS Number: 5518-52-5

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 61 to 64 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR