Triphenylphosphine dibromide (∼33% Br)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dibromotriphenylphosphorane

29107-0050 29107-0250 29107-1000
TS29107-0050EA 35 USD
TS29107-0050 TS29107-0250 TS29107-1000
Triphenylphosphine dibromide (∼33% Br)
Triphenylphosphine dibromide
Formula: C₁₈H₁₅Br₂P
MW: 422.1 g/mol
Melting Pt: 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000054
CAS Number: 1034-39-5
EINECS: 213-855-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to light pink Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ca. 33% Br2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR