1,4,8,11-Tetraazacyclotetradecane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
26933-0010
26933-0050
26933-0250
TS26933-0010EA
73
USD
TS26933-0010
TS26933-0050
TS26933-0250
1,4,8,11-Tetraazacyclotetradecane 98%
1,4,8,11-Tetraazacyclotetradecane
Formula:
C₁₀H₂₄N₄ MW: 200.33 g/mol Melting Pt: 185…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005105 CAS Number: 295-37-4 EINECS: 206-039-1 |
Specification Test Results
Appearance | White to off-white Powder or needles |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...