Triphenylphosphonium bromide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triphenylphosphine hydrobromide

26909-0050 26909-0250 26909-1000
TS26909-0050EA 30 USD
TS26909-0050 TS26909-0250 TS26909-1000
Triphenylphosphonium bromide 99%
Triphenylphosphonium bromide
Formula: C₁₈H₁₆BrP
MW: 343.2 g/mol
Melting Pt: 196…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00035107
CAS Number: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR