Benzyltrimethylammonium dichloroiodate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
26860-0050
26860-0250
26860-1000
TS26860-0050EA
49
USD
TS26860-0050
TS26860-0250
TS26860-1000
Benzyltrimethylammonium dichloroiodate 99%
Benzyltrimethylammonium dichloroiodate
Formula:
C₁₀H₁₆Cl₂IN MW: 348.06 g/mol Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075259 CAS Number: 114971-52-7 |
Specification Test Results
Appearance | Yellow to light brown Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 125 to 128 °C |
Titration with HClO4 | ≥98.5 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...