(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl

26554-0010 26554-0050 26554-2500
TS26554-0010EA 82 USD
TS26554-0010 TS26554-0050 TS26554-2500
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99+%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 238 to 242 °C
Assay ≥99.0 %
Specific optical rotation -226° to -246° (20 °C, 589 nm) (c=0.3, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR