2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-4,6-dimethoxy-s-triazine

26300-0050 26300-0250 26300-1000
TS26300-0050EA 44 USD
TS26300-0050 TS26300-0250 TS26300-1000
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Formula: C₅H₆ClN₃O₂
MW: 175.57 g/mol
Boiling Pt: 99…100 °C (1 mmHg)
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00075607
CAS Number: 3140-73-6
EINECS: 221-541-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR