γ-Cyclodextrine hydrate 99%
Supplier: Thermo Scientific Chemicals
22990-0010
22990-0050
22990-5000
TS22990-0010EA
277
USD
TS22990-0010
TS22990-0050
TS22990-5000
γ-Cyclodextrine hydrate 99%
γ-Cyclodextrine hydrate
Formula:
C₄₈H₈₀O₄₀·H₂O Melting Pt: 267 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149574 CAS Number: 91464-90-3 EINECS: 241-482-4 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤11 % |
HPLC | ≥98.5 % (ELSD) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...