Bromocresol purple sodium salt, water soluble, pure indicator

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

22961-0050 22961-0100 22961-0250 22961-1000
TS22961-0050EA 31 USD
TS22961-0050 TS22961-0100 TS22961-0250 TS22961-1000
Bromocresol purple sodium salt, water soluble, pure indicator
Bromocresol purple sodium salt
Formula: C₂₁H₁₅Br₂NaO₅S
MW: 562.21 g/mol
Melting Pt: 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148896
CAS Number: 62625-30-3
EINECS: 263-655-3

Order Now

Specification Test Results

Appearance Red-brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Specific extinction E (1%/1cm) ≥900 (590 nm, 0.01 N NaOH)
Visual transition interval from pH 5.2 (yellow) to p 6.8 (purple)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR