Eriochrome Black T, pure indicator grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T

22836-0010 22836-0250 22836-1000 22836-5000
TS22836-0010EA 182 USD
TS22836-0010 TS22836-0250 TS22836-1000 TS22836-5000
Eriochrome Black T, pure indicator grade
Eriochrome Black T
Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
Merck Index: 13,03702

Order Now

Specification Test Results

Appearance Brown to black Powder
Infrared spectrum Conforms
UV spectrum Complies
Suitability (for compl. det. of metals) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR