Chloramine-T (sodium salt) trihydrate 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

22785-0010 22785-0050 22785-0250 22785-2500
TS22785-0010EA 204 USD
TS22785-0010 TS22785-0050 TS22785-0250 TS22785-2500
Chloramine-T (sodium salt) trihydrate 97+%
Chloramine-T (sodium salt) trihydrate
Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Melting Pt: 170…177 °C (dec.)
Flash Pt: 192 °C
Storage Temperature: Ambient
MDL Number: MFCD00149066
CAS Number: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8 III
Merck Index: 13,02085

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Titration Iodometric 97.0 to 103.0 %
Appearance of solution (5% in water) Clear solution
pH 9.0 to 11.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR