α-Cyclodextrine hydrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate

22729-0050 22729-0250 22729-1000
TS22729-0050EA 63 USD
TS22729-0050 TS22729-0250 TS22729-1000
α-Cyclodextrine hydrate 98+%
α-Cyclodextrine hydrate
Formula: C₃₆H₆₀O₃₀·xH₂O
MW: 972.86 g/mol (anhy)
Melting Pt: 278 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150804
CAS Number: 51211-51-9
EINECS: 233-007-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤11 %
Sulfated ash ≤0.1 %
HPLC ≤0.5 % Linear oligomers
Specific optical rotation +145° to +151° (20°C, 589 nm) (c=2, water) (on dry substance)
Heavy metals (as Pb) ≤15 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR