Pyridinium chloride 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl

22228-0025 22228-1000 22228-5000
TS22228-0025EA 503 USD
TS22228-0025 TS22228-1000 TS22228-5000
Pyridinium chloride 98%, pure
Pyridinium chloride
Formula: C₅H₆ClN
Boiling Pt: 222.5 °C (1013 hPa)
Melting Pt: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7
EINECS: 211-027-4

Order Now

Specification Test Results

Appearance White to tan Crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Solubility almost colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR