2-Chloroethyl methyl ether 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methoxyethyl chloride

22177-0025 22177-1000 22177-5000
TS22177-0025EA 1225 USD
TS22177-0025 TS22177-1000 TS22177-5000
2-Chloroethyl methyl ether 98%
2-Chloroethyl methyl ether
Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 89…90 °C (760 mmHg)
Melting Pt: –55 °C
Density: 1.03 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4100 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR