Tetrapentylammonium bromide 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetraamylammonium bromide
22148-0050
22148-0250
22148-1000
TS22148-0050EA
37
USD
TS22148-0050
TS22148-0250
TS22148-1000
Tetrapentylammonium bromide 99+%
Tetrapentylammonium bromide
Formula:
C₂₀H₄₄BrN MW: 378.48 g/mol Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011856 CAS Number: 866-97-7 EINECS: 212-756-0 |
Specification Test Results
Appearance | White Powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 99 to 103 °C |
Titration Argentometric | ≥99.0 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...