Propionaldehyde 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propanal

22051-0025 22051-1000 22051-5000
TS22051-0025EA 426 USD
TS22051-0025 TS22051-1000 TS22051-5000
Propionaldehyde 99+%
Propionaldehyde
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (760 mmHg)
Melting Pt: –81 °C
Density: 0.798 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR